CAS RN 6737-11-7
IUPAC Name:
but-3-en-2-yl acetate
Synonyms:
but-3-en-2-yl acetate
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
LYWCPTCPTWCZSZ-UHFFFAOYNA-N
SMILES:
CC(OC(C)=O)C=C
1
–
1
of
1
results
1
3-Buten-2-yl Acetate 98.0+%, TCI America™
CAS: 6737-11-7 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.14 MDL Number: MFCD00026202 InChI Key: LYWCPTCPTWCZSZ-UHFFFAOYNA-N Synonym: 3-Acetoxy-1-butene, Acetic Acid 3-Buten-2-yl Ester, 1-Methylallyl Acetate PubChem CID: 138821 IUPAC Name: but-3-en-2-yl acetate SMILES: CC(C=C)OC(=O)C