accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONHHS

CAS RN 67385-09-5

IUPAC Name: tert-butyl N-(2-sulfanylethyl)carbamate
Synonyms: tert-butyl N-(2-sulfanylethyl)carbamate
Molecular Weight (g/mol): 177.26
Molecular Formula: C7H15NO2S
InChi Key: GSJJCZSHYJNRPN-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCS
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)