accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3C(E)

CAS RN 674-76-0

IUPAC Name: (2E)-4-methylpent-2-ene
Synonyms: trans-4-methyl-2-pentene 4-methyl-2-pentene (cis)
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: LGAQJENWWYGFSN-SNAWJCMRSA-N
SMILES: C\C=C\C(C)C
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results