accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6740-85-8

IUPAC Name: (2-chlorophenyl)(cyclopentyl)methanone
Synonyms: (2-chlorophenyl)(cyclopentyl)methanone
Molecular Weight (g/mol): 208.69
Molecular Formula: C12H13ClO
InChi Key: QIJMMRNZBJHXRI-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(=O)C1CCCC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)