accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6742-54-7

IUPAC Name: undecylbenzene
Synonyms: 1-phenylundecane
Molecular Weight (g/mol): 232.41
Molecular Formula: C17H28
InChi Key: XBEADGFTLHRJRB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results