accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SHNOOOHO

CAS RN 67436-13-9

IUPAC Name: 3-(tert-butylsulfanyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-(tert-butylsulfanyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 399.51
Molecular Formula: C22H25NO4S
InChi Key: IXAYZHCPEYTWHW-UHFFFAOYNA-N
SMILES: CC(C)(C)SCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)