CAS RN 674792-06-4
CAS RN 674792-06-4
IUPAC Name:
tert-butyl (2S)-2-(2-methylpropyl)piperazine-1-carboxylate
Synonyms:
tert-butyl (2S)-2-(2-methylpropyl)piperazine-1-carboxylate
Molecular Weight (g/mol):
242.36
Molecular Formula:
C13H26N2O2
InChi Key:
WXGGVOBNOVOVAM-NSHDSACASA-N
SMILES:
CC(C)C[C@H]1CNCCN1C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Thermo Scientific Chemicals (S)-1-Boc-2-isobutylpiperazine, 97%
CAS: 674792-06-4 Molecular Formula: C13H26N2O2 Molecular Weight (g/mol): 242.363 MDL Number: MFCD03787916 InChI Key: WXGGVOBNOVOVAM-NSHDSACASA-N Synonym: s-1-boc-2-isobutyl-piperazine,s-tert-butyl 2-isobutylpiperazine-1-carboxylate,s-1-boc-2-isobutylpiperazine,tert-butyl 2s-2-2-methylpropyl piperazine-1-carboxylate,s-1-n-boc-isobutylpiperazine,s-n-boc-2-isobutylpiperazine,tert-butyl 2s-2-isobutylpiperazine-1-carboxylate,s-1-n-boc-2-isobutylpiperazine,2s-1-n-boc-2-isobutylpiperazine,2s-1-tert-butoxycarbonyl-2-isobutylpiperazine PubChem CID: 17750437 IUPAC Name: tert-butyl (2S)-2-(2-methylpropyl)piperazine-1-carboxylate SMILES: CC(C)CC1CNCCN1C(=O)OC(C)(C)C