accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOOO

CAS RN 6748-68-1

IUPAC Name: 8-acetyl-7-hydroxy-2H-chromen-2-one
Synonyms: 8-acetyl-7-hydroxychromen-2-one
Molecular Weight (g/mol): 204.18
Molecular Formula: C11H8O4
InChi Key: XWYMACPLPPQCHC-UHFFFAOYSA-N
SMILES: CC(=O)C1=C2OC(=O)C=CC2=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results