accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 675-20-7

IUPAC Name: piperidin-2-one
Synonyms: 2-piperidinone delta-valerolactam
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: XUWHAWMETYGRKB-UHFFFAOYSA-N
SMILES: O=C1CCCCN1
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
Quantity 
  • (4)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)