accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 675-62-7

IUPAC Name: dichloro(methyl)(3,3,3-trifluoropropyl)silane
Synonyms: dichloro(methyl)(3,3,3-trifluoropropyl)silane
Molecular Weight (g/mol): 211.08
Molecular Formula: C4H7Cl2F3Si
InChi Key: OHABWQNEJUUFAV-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)CCC(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results