accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CCH3H3CMnCCH3CCH3CCH3CH3CCCH3H3CH3C

CAS RN 67506-86-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 325.4
Molecular Formula: C20H30Mn
InChi Key: DBNMEMKBUCQRTO-UHFFFAOYSA-N
SMILES: Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Mn]2345

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results