accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67515-59-7

IUPAC Name: 4-fluoro-3-(trifluoromethyl)benzonitrile
Synonyms: 4-fluoro-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 189.11
Molecular Formula: C8H3F4N
InChi Key: CQZQCORFYSSCFY-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1C(F)(F)F)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results