accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67515-60-0

FOFFF

CAS RN 67515-60-0

IUPAC Name: 4-fluoro-3-(trifluoromethyl)benzaldehyde
Synonyms: 4-fluoro-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: BIUDHHGROGJSHN-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C=C1C(F)(F)F
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results