CAS RN 6753-62-4
IUPAC Name:
2-(methylamino)pentanedioic acid hydrochloride
Synonyms:
N-methyl-L-glutamic acid
Molecular Weight (g/mol):
197.62
Molecular Formula:
C6H12ClNO4
InChi Key:
LAYOOVKHWQMRIQ-UHFFFAOYNA-N
SMILES:
Cl.CNC(CCC(O)=O)C(O)=O
1
–
1
of
1
results
1
N-methyl-L-glutamic acid, MP Biomedicals™
CAS: 6753-62-4 Molecular Formula: C6H12ClNO4 Molecular Weight (g/mol): 197.615 MDL Number: MFCD00065920 InChI Key: LAYOOVKHWQMRIQ-WCCKRBBISA-N Synonym: n-me-glu-oh,h-l-meglu-oh hcl,n-alpha-methyl-l-glutamic acid hydrochloride,n-methyl-l-glutamic acid hydrochloride,s-2-methylamino pentanedioic acid hydrochloride PubChem CID: 56777325 IUPAC Name: (2S)-2-(methylamino)pentanedioic acid;hydrochloride SMILES: CNC(CCC(=O)O)C(=O)O.Cl