accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3CH3(E)(E)(E)

CAS RN 6753-98-6

IUPAC Name: (1E,4E,8E)-2,6,6,9-tetramethylcycloundeca-1,4,8-triene
Synonyms: humulene 3,7,10-humulatriene (1E,4E,8E)-α-humulene α-humulene α-caryophyllene
Molecular Weight (g/mol): 204.36
Molecular Formula: C15H24
InChi Key: FAMPSKZZVDUYOS-HRGUGZIWSA-N
SMILES: C\C1=C/CC(C)(C)\C=C\C\C(C)=C\CC1
Grade 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)