accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67536-29-2

IUPAC Name: 1-methoxy-1,3-dihydro-2-benzofuran
Synonyms: 1-methoxy-1,3-dihydro-2-benzofuran
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: HHFOTZJVYJNYJC-UHFFFAOYNA-N
SMILES: COC1OCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results