accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67580-39-6

IUPAC Name: sodium 9,10-dimethoxyanthracene-2-sulfonate
Synonyms: sodium 9,10-dimethoxyanthracene-2-sulfonate
Molecular Weight (g/mol): 340.32
Molecular Formula: C16H13NaO5S
InChi Key: CBGQZXMEEWCJQD-UHFFFAOYSA-M
SMILES: [Na+].COC1=C2C=CC(=CC2=C(OC)C2=CC=CC=C12)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results