accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNF

CAS RN 675831-63-7

IUPAC Name: 3-(5-fluoro-1H-indol-3-yl)cyclopentan-1-one
Synonyms: 3-(5-fluoro-1H-indol-3-yl)cyclopentan-1-one
Molecular Weight (g/mol): 217.24
Molecular Formula: C13H12FNO
InChi Key: DINNCXVUOXLHSH-UHFFFAOYNA-N
SMILES: FC1=CC=C2NC=C(C3CCC(=O)C3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results