accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SCH3H3CFBFFF

CAS RN 676-88-0

IUPAC Name: tetrafluoroboranuide; trimethylsulfanium
Synonyms: trimethylsulfonium tetrafluoroborate
Molecular Weight (g/mol): 163.97
Molecular Formula: C3H9BF4S
InChi Key: RNBNWTAKWJKQNN-UHFFFAOYSA-N
SMILES: C[S+](C)C.F[B-](F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)