accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 676-96-0

IUPAC Name: (dimethylphosphoryl)methane
Synonyms: (dimethylphosphoryl)methane
Molecular Weight (g/mol): 92.08
Molecular Formula: C3H9OP
InChi Key: LRMLWYXJORUTBG-UHFFFAOYSA-N
SMILES: CP(C)(C)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results