accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SHCH3O

CAS RN 67633-97-0

IUPAC Name: 3-sulfanylpentan-2-one
Synonyms: 3-sulfanylpentan-2-one
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: SZECUQRKLXRGSJ-UHFFFAOYNA-N
SMILES: CCC(S)C(C)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results