accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67639-41-2

IUPAC Name: 4-azaniumylthiane-4-carboxylate
Synonyms: 4-ammoniothiane-4-carboxylate
Molecular Weight (g/mol): 161.22
Molecular Formula: C6H11NO2S
InChi Key: WIPRZHGCPZSPLJ-UHFFFAOYSA-N
SMILES: [NH3+]C1(CCSCC1)C([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results