CAS RN 67655-94-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Amastatin, Streptomyces sp.
Amastatin is a non-toxic inhibitor of aminopeptidase A (Ki = 1 μM) and leucine aminopeptidase.