accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67715-80-4

IUPAC Name: 2-methyl-4-propyl-1,3-oxathiane
Synonyms: 2-methyl-4-propyl-1,3-oxathiane
Molecular Weight (g/mol): 160.28
Molecular Formula: C8H16OS
InChi Key: GKGOLPMYJJXRGD-UHFFFAOYNA-N
SMILES: CCCC1CCOC(C)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results