accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67719-69-1

IUPAC Name: titanium(4+) (dimethylalumanyl)methanide bis(cyclopenta-2,4-dien-1-ide) chloride
Synonyms: tebbe's reagent
Molecular Weight (g/mol): 284.59
Molecular Formula: C13H18AlClTi
InChi Key: HIVIAXIRSNAVAD-UHFFFAOYSA-M
SMILES: [Cl-].[Ti+4].C[Al](C)[CH2-].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)