accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6775-01-5

IUPAC Name: bis(3-aminopropyl)(phenyl)phosphane
Synonyms: bis(3-aminopropyl)(phenyl)phosphane
Molecular Weight (g/mol): 224.29
Molecular Formula: C12H21N2P
InChi Key: OEHBMEXOPYWPKQ-UHFFFAOYSA-N
SMILES: NCCCP(CCCN)C1=CC=CC=C1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results