accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COOCH3OCH3(E/Z)

CAS RN 67751-23-9

IUPAC Name: (4,4-dimethoxy-3-oxobut-1-en-1-yl)dimethylamine
Synonyms: (4,4-dimethoxy-3-oxobut-1-en-1-yl)dimethylamine
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: DFZIBCAWOSFLFR-UHFFFAOYSA-N
SMILES: COC(OC)C(=O)C=CN(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results