accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNON

CAS RN 6777-05-5

IUPAC Name: 1-benzylimidazolidine-2,4-dione
Synonyms: N-benzyl-2,4-imidazolidione
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: VJUNTPRQTFDQMF-UHFFFAOYSA-N
SMILES: O=C1CN(CC2=CC=CC=C2)C(=O)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results