accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6779-09-5

IUPAC Name: ethylphosphonate
Synonyms: ethylphosphonate
Molecular Weight (g/mol): 108.03
Molecular Formula: C2H5O3P
InChi Key: GATNOFPXSDHULC-UHFFFAOYSA-L
SMILES: CCP([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results