accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFFFFFFFFFFFFFFFF

CAS RN 678-39-7

IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecan-1-ol
Synonyms: 8:2 fluorotelomer alcohol 1H,1H,2H,2H-perfluorodecanol
Molecular Weight (g/mol): 464.12
Molecular Formula: C10H5F17O
InChi Key: JJUBFBTUBACDHW-UHFFFAOYSA-N
SMILES: OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)