accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHFFFFFFFFFFFFHO

CAS RN 678-45-5

IUPAC Name: dodecafluorooctanedioic acid
Synonyms: dodecafluorooctanedioic acid
Molecular Weight (g/mol): 390.08
Molecular Formula: C8H2F12O4
InChi Key: QYCPVKMFWNBDPV-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results