accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67818-41-1

IUPAC Name: 1-(2-chloro-4-nitrophenyl)ethan-1-one
Synonyms: 1-(2-chloro-4-nitrophenyl)ethanone
Molecular Weight (g/mol): 199.59
Molecular Formula: C8H6ClNO3
InChi Key: BRFBJVQQCATLSZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results