accessibility menu, dialog, popup

UserName

Viewing profile as user:
BCH3H3CH3CBCH3H3CH3CH3CNCH3NCH3CH3

CAS RN 67826-92-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 412.32
Molecular Formula: C26H50B2N2
InChi Key: YIPGSNPACSKQKJ-UHFFFAOYNA-N
SMILES: CN(C)CCN(C)C.[B]C1CC2CC(C1C)C2(C)C.[B]C1CC2CC(C1C)C2(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results