accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrN

CAS RN 67832-11-5

IUPAC Name: 4-bromo-2-methylbenzonitrile
Synonyms: 4-bromo-2-methylbenzonitrile
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: LPEBMDFRIKYFCF-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results