CAS RN 67884-32-6
IUPAC Name:
[1-(diphenylphosphanyl)propan-2-yl]diphenylphosphane
Synonyms:
[1-(diphenylphosphanyl)propan-2-yl]diphenylphosphane
Molecular Weight (g/mol):
412.45
Molecular Formula:
C27H26P2
InChi Key:
WGOBPPNNYVSJTE-UHFFFAOYNA-N
SMILES:
CC(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich Magnesium Bis(Monoperoxyphthalate) Hexahydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.