accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67939-81-5

IUPAC Name: tris[(2-ethylhexanoyl)oxy]silyl 2-ethylhexanoate
Synonyms: tris[(2-ethylhexanoyl)oxy]silyl 2-ethylhexanoate
Molecular Weight (g/mol): 600.91
Molecular Formula: C32H60O8Si
InChi Key: UNKOLEUDCQIVBV-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(=O)O[Si](OC(=O)C(CC)CCCC)(OC(=O)C(CC)CCCC)OC(=O)C(CC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results