accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClFI

CAS RN 6797-79-1

IUPAC Name: 4-chloro-2-fluoro-1-iodobenzene
Synonyms: 4-chloro-2-fluoro-1-iodobenzene
Molecular Weight (g/mol): 256.44
Molecular Formula: C6H3ClFI
InChi Key: RSTFBOIFYXJIMR-UHFFFAOYSA-N
SMILES: FC1=C(I)C=CC(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results