CAS RN 68-04-2
Buffer solution pH 4, Citrate buffer
CAS: 68-04-2 | C8H5KO4
Trisodium citrate, anhydrous, 99%
CAS: 68-04-2 Molecular Formula: C6H5Na3O7 Molecular Weight (g/mol): 258.07 MDL Number: MFCD00012462 InChI Key: HRXKRNGNAMMEHJ-UHFFFAOYSA-K Synonym: sodium citrate,trisodium citrate,citrosodine,sodium citrate anhydrous,citrosodina,natrocitral,citnatin,citreme,citrosodna,trisodium citrate, anhydrous PubChem CID: 6224 ChEBI: CHEBI:53258 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate SMILES: [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Sodium Citrate, Anhydrous, USP, 99-100.5%, Spectrum™ Chemical
CAS: 68-04-2 Molecular Formula: C6H5Na3O7 Molecular Weight (g/mol): 258.07 MDL Number: MFCD00012462 InChI Key: HRXKRNGNAMMEHJ-UHFFFAOYSA-K IUPAC Name: trisodium 2-hydroxypropane-1,2,3-tricarboxylate SMILES: [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Sodium Citrate, Anhydrous, Powder, FCC, 99-100.5%, Spectrum™ Chemical
CAS: 68-04-2 Molecular Formula: C6H5Na3O7 Molecular Weight (g/mol): 258.07 MDL Number: MFCD00012462 InChI Key: HRXKRNGNAMMEHJ-UHFFFAOYSA-K IUPAC Name: trisodium 2-hydroxypropane-1,2,3-tricarboxylate SMILES: [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Citric acid, trisodium salt, 98%, pure, anhydrous
CAS: 68-04-2 Molecular Formula: C6H5Na3O7 Molecular Weight (g/mol): 258.07 MDL Number: MFCD00012462 InChI Key: HRXKRNGNAMMEHJ-UHFFFAOYSA-K Synonym: sodium citrate,trisodium citrate,citrosodine,sodium citrate anhydrous,citrosodina,natrocitral,citnatin,citreme,citrosodna,trisodium citrate, anhydrous PubChem CID: 6224 ChEBI: CHEBI:53258 IUPAC Name: trisodium;2-hydroxypropane-1,2,3-tricarboxylate SMILES: [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O