accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 680569-83-9

IUPAC Name: 6-chloro-1-methyl-1H-indole-2-carboxylic acid
Synonyms: 6-chloro-1-methylindole-2-carboxylic acid
Molecular Weight (g/mol): 209.63
Molecular Formula: C10H8ClNO2
InChi Key: BMHTUBVXMTYFAS-UHFFFAOYSA-N
SMILES: CN1C(=CC2=CC=C(Cl)C=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results