accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 6812-38-0

IUPAC Name: hexadecylcyclohexane
Synonyms: 1-cyclohexylhexadecane
Molecular Weight (g/mol): 308.59
Molecular Formula: C22H44
InChi Key: NRHBFNBZAZZTAU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results