CAS RN 68121-46-0
CAS RN 68121-46-0
IUPAC Name:
1,3-dichloro-5-(methylsulfanyl)benzene
Synonyms:
1,3-dichloro-5-(methylsulfanyl)benzene
Molecular Weight (g/mol):
193.09
Molecular Formula:
C7H6Cl2S
InChi Key:
PSSCMQMQOXVMFG-UHFFFAOYSA-N
SMILES:
CSC1=CC(Cl)=CC(Cl)=C1
1
–
1
of
1
results
1
3,5-Dichlorothioanisole 98.0+%, TCI America™
CAS: 68121-46-0 Molecular Formula: C7H6Cl2S Molecular Weight (g/mol): 193.09 MDL Number: MFCD00870149 InChI Key: PSSCMQMQOXVMFG-UHFFFAOYSA-N Synonym: 1,3-Dichloro-5-(methylthio)benzene PubChem CID: 129278 IUPAC Name: 1,3-dichloro-5-(methylsulfanyl)benzene SMILES: CSC1=CC(Cl)=CC(Cl)=C1