accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68123-33-1

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-(4-nitrophenyl)acetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-(4-nitrophenyl)acetate
Molecular Weight (g/mol): 278.22
Molecular Formula: C12H10N2O6
InChi Key: JUEAHIVORWVODA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CC(=O)ON2C(=O)CCC2=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results