accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COO∗n

CAS RN 68131-40-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: [C2H4O]n C13H27O
InChi Key: —
SMILES: CCCCCCCC(CCCCC)OCCO-*
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results