accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4NH4ClClClClClRu

CAS RN 68133-88-0

IUPAC Name: ruthenium(3+) diammonium pentachloride
Synonyms: ruthenium(3+) diammonium pentachloride
Molecular Weight (g/mol): 314.4
Molecular Formula: Cl5H8N2Ru
InChi Key: AEETWQIZFGAMIM-UHFFFAOYSA-K
SMILES: [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results