accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 681435-25-6

IUPAC Name: cadmium(2+) dibromide
Synonyms: cadmium bromide
Molecular Weight (g/mol): 272.22
Molecular Formula: Br2Cd
InChi Key: KPWJBEFBFLRCLH-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Cd++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results