accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBrBrOH

CAS RN 681481-91-4

IUPAC Name: 4,4'-dibromo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol
Synonyms: 4,4'-dibromo-2,2',3,3'-tetrahydro-1,1'-spirobi[indene]-7,7'-diol
Molecular Weight (g/mol): 410.11
Molecular Formula: C17H14Br2O2
InChi Key: OZVMVFKUNSNXQC-UHFFFAOYSA-N
SMILES: OC1=C2C(CCC22CCC3=C(Br)C=CC(O)=C23)=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results