accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 681508-91-8

IUPAC Name: tert-butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate
Synonyms: tert-butyl 4-{4-[(methylamino)methyl]phenyl}piperazine-1-carboxylate
Molecular Weight (g/mol): 305.42
Molecular Formula: C17H27N3O2
InChi Key: VERAVZPFPZKVCT-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)N1CCN(CC1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results