accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68176-57-8

IUPAC Name: 4-tert-butylbenzene-1,2-diamine
Synonyms: 4-tert-butylbenzene-1,2-diamine
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: WLOSFXSXVXTKBU-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(N)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results