accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOOHHONH2HOOHHOHOPHOOONOO

CAS RN 68189-42-4

IUPAC Name: (4-nitrophenoxy)phosphonic acid; bis(2-amino-2-(hydroxymethyl)propane-1,3-diol)
Synonyms: 4-nitrophenyl phosphate; bis(tris buffer)
Molecular Weight (g/mol): 461.36
Molecular Formula: C14H28N3O12P
InChi Key: XXAXKCWOTRABOW-UHFFFAOYSA-N
SMILES: NC(CO)(CO)CO.NC(CO)(CO)CO.OP(O)(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results