accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 682-30-4

IUPAC Name: diethyl ethenylphosphonate
Synonyms: diethyl ethenylphosphonate
Molecular Weight (g/mol): 164.14
Molecular Formula: C6H13O3P
InChi Key: DREPONDJUKIQLX-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C=C
Boiling Point 
  • (4)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)